Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HO-1/HMOX1/HSP32 Antibody (6B8-2F2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259338F
Description
HO-1/HMOX1/HSP32 Monoclonal specifically detects HO-1/HMOX1/HSP32 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
HO-1/HMOX1/HSP32 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
HMOX1 | |
His-tagged Rat HO-1 | |
100 μL | |
Primary | |
Detects 32kDa. Does not cross-react with HO-2. | |
Store at 4C in the dark. | |
IgG1 κ |
Immunocytochemistry, Immunofluorescence | |
6B8-2F2 | |
Immunocytochemistry/Immunofluorescence | |
bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32 | |
Mouse | |
Protein G purified | |
Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism | |
3162 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction