Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HO-1/HMOX1/HSP32 Antibody (6B8-2F2), Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | HO-1/HMOX1/HSP32 |
---|---|
Clone | 6B8-2F2 |
Dilution | Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
HO-1/HMOX1/HSP32 Monoclonal specifically detects HO-1/HMOX1/HSP32 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
HO-1/HMOX1/HSP32 | |
Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
3162 | |
His-tagged Rat HO-1 | |
Primary | |
Detects 32kDa. Does not cross-react with HO-2. |
6B8-2F2 | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism | |
bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32 | |
HMOX1 | |
IgG1 κ | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title