Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HO-1/HMOX1/HSP32 Mouse anti-Human, Biotin, Clone: 1D6, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259492B
This item is not returnable.
View return policy
Description
HO-1/HMOX1/HSP32 Monoclonal antibody specifically detects HO-1/HMOX1/HSP32 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
HO-1/HMOX1/HSP32 | |
Monoclonal | |
Biotin | |
bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32 | |
Recombinant human HO-1/HMOX1/HSP32 (1-266aa) purified from E. coli | |
0.1 mL | |
Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism | |
3162 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
1D6 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction