Learn More
Specifications
Specifications
| For Use With (Application) | SDS-PAGE, Western Blot |
| Formulation | 20mM Tris pH7.5, 0.1mM EDTA, 0.1% triton x-100, 0.2% Na cholate, <50mM KCl |
| Gene ID (Entrez) | 3162 |
| Molecular Weight (g/mol) | M.W. Theoretical: 33 kDa |
| Quantity | 50 μg |
| Storage Requirements | Store at -70C. Avoid freeze-thaw cycles. |
| Gene Alias | bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32 |
| Gene Symbol | HMOX1 |
| Research Category | Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism |
| Purity or Quality Grade | >85% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
