Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HO-2/HMOX2 Antibody (OTI1C2), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270895FR
Description
HO-2/HMOX2 Monoclonal specifically detects HO-2/HMOX2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
HO-2/HMOX2 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
HMOX2 | |
Full length recombinant protein of human HO-2/HMOX2 (NP_002125) produced in HEK293T cells. | |
0.1 mL | |
3163.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI1C2 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
heme oxygenase (decycling) 2, heme oxygenase 2, HO2, HO-2EC 1.14.99.3 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction