Learn More
Eisco™ Hollow Glass Prisms
SCI_ED
Catalog No. S41520 Shop All Eisco Products
Change view
Click to view available options
Product Type:
Hollow Cube
Hollow Prism Cube
Hollow Pyramidal
Hollow Rectangular, Slab
Length (Metric):
2.5 cm
8.29 cm
8.9 cm
10.2 cm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Length (Metric)
S41520 Hollow Pyramidal 8.29 cm
S27411 Hollow Prism Cube 2.5 cm
S41526 Hollow Cube 10.2 cm
S41521 Hollow Pyramidal 10.2 cm
S41524 Hollow Rectangular, Slab 8.9 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S41520 Supplier Eisco™ Supplier No. PH0552A
Add to Cart
Add to Cart
Includes
prism with stopper
Hollow glass prisms are made from glass plates that are cemented together. Stoppers included. Excellent tools for optical experimentation as well as for studying Snells Law of Refraction. Change the liquids or gases in the prisms to change the refraction properties
  • Made from glass plates, cemented together.
  • Stopper included
  • Hollow inside allows for a variety of optic experiments
  • Excellent tool for studying Snells Law of Refraction.
  • Designed for educational use
Warranty and Services
1 year limited manufacturer warranty
Specifications
Product Type Hollow Pyramidal
Material Glass
For Use With (Application) Optics experiments, Snell's Law of Refraction
Includes Prism, Stopper
Quantity 1
Length (Metric) 8.29 cm
Weight (Metric) 50 g