Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Holmium(III) acetate hydrate, REacton™, 99.9% (REO)

Catalog No. AA1459109
Click to view available options
Quantity:
2 g
10 g
50 g

CAS: 312619-49-1 | C6H9HoO6 | 342.06 g/mol

Holmium(III) acetate hydrate is used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312619-49-1
Molecular Formula C6H9HoO6
Molecular Weight (g/mol) 342.06
MDL Number MFCD00150648
InChI Key HWQRVFRWLQGBFV-UHFFFAOYSA-K
Synonym holmium iii acetate hexahydrate, holmium iii acetate hydrate, reacton reo, holmium iii acetatehexahydrate, holmium acetate-water 1/3/6
IUPAC Name holmium(3+) triacetate
SMILES [Ho+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 10 g
Solubility Information Soluble in water.
Formula Weight 342.07 (Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Assay (REO)
Chemical Name or Material Holmium(III) acetate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 247-066-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.