Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Holmium(III) oxide, 99.995% (REO), Alfa Aesar™, Reacton™

Manufacturer:  Alfa Aesar 1091014

Quantity 25g

 View more versions of this product

Catalog No. AA1091014


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12055-62-8
99.995% (REO)
377.86
Holmium(III) oxide
Powder or Lump
Ho2O3
MFCD00011053

Specifications

12055-62-8
99.995% (REO)
Beige powder or lump
MFCD00011053
Beige
Holmium(III) oxide
Powder or Lump
Ho2O3
377.86
Odorless
25g

Safety and Handling

SDS & Safety Documents

EINECSNumber : 235-015-3

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit