Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOMER1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299478APC
Description
HOMER1 Polyclonal antibody specifically detects HOMER1 in Human samples. It is validated for Western BlotSpecifications
HOMER1 | |
Polyclonal | |
Western Blot | |
HOMER, homer homolog 1 (Drosophila), homer-1, HOMER1B, HOMER-1B, HOMER1C, SYN47neuronal immediate early gene, 1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HOMER1. | |
0.1 mL | |
Neuroscience, Neurotransmission | |
9456 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction