Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOMER1 Antibody (SR1770), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32260050UL
Description
HOMER1 Monoclonal antibody specifically detects HOMER1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
HOMER1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
SR1770 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
HOMER, homer homolog 1 (Drosophila), homer-1, HOMER1B, HOMER-1B, HOMER1C, SYN47neuronal immediate early gene, 1 | |
A synthesized peptide derived from human HOMER1 (Uniprot #: Q86YM7) | |
50 μg | |
Neuroscience, Neurotransmission | |
9456 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction