Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOMER1 Monoclonal Antibody (OTI3G5), TrueMAB™, OriGene
SDP

Catalog No. 502543936
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3936 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3936 Supplier OriGene Supplier No. CF811741
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HOMER1 Monoclonal antibody specifically detects HOMER1 in Human samples. It is validated for Western Blot

Specifications

Antigen HOMER1
Applications Western Blot
Classification Monoclonal
Clone OTI3G5
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene BAG5
Gene Accession No. Q9UL15
Gene Alias HOMER; homer homolog 1; homer homolog 1 (Drosophila); homer protein homolog 1; homer scaffolding protein 1; homer, neuronal immediate early gene, 1; Homer1; homer-1; HOMER1A; HOMER1B; HOMER1C; HOMER1F; immediate early gene protein Homer1A; PSD-Zip45; scaffold protein; SYN47; VASP/Ena-related gene up-regulated during seizure and LTP 1; Ves-1; Vesl; Vesl1; Vesl-1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 290-440 of human BAG5 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9529
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.