Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOMER1 Mouse anti-Human, Mouse, Rat, Clone: OTI7G10, lyophilized, TrueMAB™
SDP

Catalog No. 501670820
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-0820 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-0820 Supplier OriGene Supplier No. CF811745
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Homer 1 (Homer protein homolog 1) belongs to the Homer family of proteins that regulate signal transduction through, and the trafficking of, glutamate receptors, as well as maintain and regulate extracellular glutamate levels in corticolimbic brain regions. The Homer 1 is enriched at excitatory synapses and binds group 1 metabotropic glutamate receptors (mGluRs).

Specifications

Antigen HOMER1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7G10
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene HOMER1
Gene Accession No. Q86YM7, Q9Z214, Q9Z2Y3
Gene Alias HOMER, HOMER1A, HOMER1B, HOMER1C, SYN47, Ves-1
Gene Symbols HOMER1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-354 of human HOMER1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26556, 29546, 9456
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.