Learn More
Invitrogen™ HOMER1 Polyclonal Antibody

Description
The target sequence has 99% sequence homology with rat, 98% to bovine, 96% to xenopus and 74% to zebrafish.
Specifications
Specifications
| Antigen | HOMER1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with 0.1% sodium azide |
| Gene | Homer1 |
| Gene Accession No. | Q86YM7, Q9Z2Y3 |
| Gene Alias | HOMER; homer homolog 1; homer homolog 1 (Drosophila); homer protein homolog 1; homer scaffolding protein 1; homer, neuronal immediate early gene, 1; Homer1; homer-1; HOMER1A; HOMER1B; HOMER1C; HOMER1F; immediate early gene protein Homer1A; PSD-Zip45; scaffold protein; SYN47; VASP/Ena-related gene up-regulated during seizure and LTP 1; Ves-1; Vesl; Vesl1; Vesl-1 |
| Gene Symbols | Homer1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.