Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOMER1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 124331AP150UL
Description
HOMER1 is a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function.Specifications
| HOMER1 | |
| Polyclonal | |
| Unconjugated | |
| HOMER1 | |
| HOMER, Homer 1, homer homolog 1 (Drosophila), Homer protein homolog 1, HOMER1, HOMER1A, HOMER1B, HOMER1C, SYN47, Ves 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26556, 29546, 9456 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.46 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86YM7, Q9Z214, Q9Z2Y3 | |
| HOMER1 | |
| HOMER1 Fusion Protein Ag3109 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction