Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOMER2 Antibody (OTI1D1), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271464H
Description
HOMER2 Monoclonal specifically detects HOMER2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
HOMER2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
HOMER2 | |
Full length recombinant protein of human HOMER2 (NP_004830) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot | |
OTI1D1 | |
Western Blot | |
ACPD, CPD, cupidin, homer homolog 2 (Drosophila), homer homolog 3, homer protein homolog 2, homer, neuronal immediate early gene, 2, Homer-2, HOMER2A, HOMER-2A, HOMER2B, HOMER-2B, Vesl-2 | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
9455.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction