Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOMER2 Antibody (OTI1H3), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271465AF532
Description
HOMER2 Monoclonal specifically detects HOMER2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
HOMER2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
HOMER2 | |
Full length recombinant protein of human HOMER2 (NP_004830) produced in E.coli. | |
DNA replication Transcription Translation and Splicing | |
9455 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1H3 | |
Western Blot | |
ACPD, CPD, cupidin, homer homolog 2 (Drosophila), homer homolog 3, homer protein homolog 2, homer, neuronal immediate early gene, 2, Homer-2, HOMER2A, HOMER-2A, HOMER2B, HOMER-2B, Vesl-2 | |
Mouse | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction