Learn More
Invitrogen™ HOMER2 Polyclonal Antibody

Description
The antibody detects endogenous levels of total HOMER2 protein.
Specifications
Specifications
| Antigen | HOMER2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | HOMER2 |
| Gene Accession No. | Q9NSB8 |
| Gene Alias | 9330120H11Rik; A VESL-2; ACPD; AW539445; CPD; Cupidin; DFNA68; homer 2; homer homolog 2; homer homolog 2 (Drosophila); homer homolog 3; homer protein homolog 2; homer scaffold protein 2; homer scaffolding protein 2; homer, neuronal immediate early gene, 2; HOMER2; homer-2; VASP/Ena-related gene up-regulated during seizure and LTP 2; Vesl2; Vesl-2 |
| Gene Symbols | HOMER2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.