Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOMER2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559078 Supplier Invitrogen™ Supplier No. PA559078
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IIPQLMSECE YVSEKLEAAE RDNQNLEDKV RSLKTDIEES KYRQRHLKVE LKSFLEVLDG KIDDLHDFRR GLSKLGTDN Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene encodes a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function. The encoded protein is a postsynaptic density scaffolding protein. Alternative splicing results in multiple transcript variants. Two related pseudogenes have been identified on chromosome 14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOMER2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HOMER2
Gene Accession No. Q9NSB8
Gene Alias 9330120H11Rik; A VESL-2; ACPD; AW539445; CPD; Cupidin; DFNA68; homer 2; homer homolog 2; homer homolog 2 (Drosophila); homer homolog 3; homer protein homolog 2; homer scaffold protein 2; homer scaffolding protein 2; homer, neuronal immediate early gene, 2; HOMER2; homer-2; VASP/Ena-related gene up-regulated during seizure and LTP 2; Vesl2; Vesl-2
Gene Symbols HOMER2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HOMER2. Recombinant protein control fragment (Product #RP-98050).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9455
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.