Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOMER2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570541
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570541 Supplier Invitrogen™ Supplier No. PA570541
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.

This gene encodes a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function. The encoded protein is a postsynaptic density scaffolding protein. Alternative splicing results in multiple transcript variants. Two related pseudogenes have been identified on chromosome 14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOMER2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HOMER2
Gene Accession No. Q9QWW1
Gene Alias 9330120H11Rik; A VESL-2; ACPD; AW539445; CPD; Cupidin; DFNA68; homer 2; homer homolog 2; homer homolog 2 (Drosophila); homer homolog 3; homer protein homolog 2; homer scaffold protein 2; homer scaffolding protein 2; homer, neuronal immediate early gene, 2; HOMER2; homer-2; VASP/Ena-related gene up-regulated during seizure and LTP 2; Vesl2; Vesl-2
Gene Symbols HOMER2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse HOMER2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26557
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.