Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HOMER3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559383
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559383 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559383 Supplier Invitrogen™ Supplier No. PA559383
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALVQTKDQEI QTLKSQTGGP REALEAAERE ETQQKVQDLE TRNAELEHQL RAMERSLEEA RAERERARAE VGRAAQLLDV RLFELSELRE GL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.

This gene encodes a member of the HOMER family of postsynaptic density scaffolding proteins that share a similar domain structure consisting of an N-terminal Enabled/vasodilator-stimulated phosphoprotein homology 1 domain which mediates protein-protein interactions, and a carboxy-terminal coiled-coil domain and two leucine zipper motifs that are involved in self-oligomerization. The encoded protein binds numerous other proteins including group I metabotropic glutamate receptors, inositol 1,4,5-trisphosphate receptors and amyloid precursor proteins and has been implicated in diverse biological functions such as neuronal signaling, T-cell activation and trafficking of amyloid beta peptides. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HOMER3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HOMER3
Gene Accession No. Q9NSC5
Gene Alias AW146114; homer homolog 3; homer homolog 3 (Drosophila); homer protein homolog 3; homer scaffolding protein 3; Homer, neuronal immediate early gene, 3; HOMER3; homer-3; VASP/Ena-related gene up-regulated during seizure and LTP 3; Vesl3; Vesl-3
Gene Symbols HOMER3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HOMER3. Recombinant protein control fragment (Product #RP-98314).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9454
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.