Learn More
Invitrogen™ HOMER3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579384
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human PC-3 whole cell, human U251 whole cell, human A431 whole cell, rat brain tissue, rat lung tissuemouse brain tissuemouse lung tissue. IHC: human mammary cancer tissue, human intestinal cancer tissue, human lung cancer tissue.
This gene encodes a member of the HOMER family of postsynaptic density scaffolding proteins that share a similar domain structure consisting of an N-terminal Enabled/vasodilator-stimulated phosphoprotein homology 1 domain which mediates protein-protein interactions, and a carboxy-terminal coiled-coil domain and two leucine zipper motifs that are involved in self-oligomerization. The encoded protein binds numerous other proteins including group I metabotropic glutamate receptors, inositol 1,4,5-trisphosphate receptors and amyloid precursor proteins and has been implicated in diverse biological functions such as neuronal signaling, T-cell activation and trafficking of amyloid beta peptides. Alternative splicing results in multiple transcript variants.
Specifications
HOMER3 | |
Polyclonal | |
Unconjugated | |
HOMER3 | |
AW146114; homer homolog 3; homer homolog 3 (Drosophila); homer protein homolog 3; homer scaffolding protein 3; Homer, neuronal immediate early gene, 3; HOMER3; homer-3; VASP/Ena-related gene up-regulated during seizure and LTP 3; Vesl3; Vesl-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26558, 29548, 9454 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
Q99JP6, Q9NSC5, Q9Z2X5 | |
HOMER3 | |
E. coli-derived human HOMER3 recombinant protein (Position: R282-A360). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.