Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Homez Antibody (PCRP-HOMEZ-1A5), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308302MFV450
Description
Homez Monoclonal antibody specifically detects Homez in Human samples. It is validated for Flow CytometrySpecifications
Homez | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
57594 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
PCRP-HOMEZ-1A5 | |
Flow Cytometry | |
homeobox and leucine zipper encoding, homeobox and leucine zipper protein Homez, KIAA1443Homeodomain leucine zipper-containing factor | |
Recombinant full-length human Homez protein (Uniprot: Q8IX15) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction