Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Homez Antibody (PCRP-HOMEZ-1B5), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308303PCP
Description
Homez Monoclonal antibody specifically detects Homez in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Protein ArraySpecifications
Homez | |
Monoclonal | |
PerCP | |
homeobox and leucine zipper encoding, homeobox and leucine zipper protein Homez, KIAA1443Homeodomain leucine zipper-containing factor | |
Recombinant full-length human Homez protein (Uniprot: Q8IX15) | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry (Paraffin), Protein Array | |
PCRP-HOMEZ-1B5 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
57594 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction