Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Homez Mouse anti-Human, Clone: PCRP-HOMEZ-1A5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30725720UG
Description
Homez Monoclonal specifically detects Homez in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Homez | |
| Monoclonal | |
| 0.2 mg/ml | |
| Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin, Protein Array | |
| homeobox and leucine zipper encoding, homeobox and leucine zipper protein Homez, KIAA1443Homeodomain leucine zipper-containing factor | |
| Recombinant full-length human Homez protein | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
| PCRP-HOMEZ-1A5 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 57594 | |
| Store at 4°C. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction