Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Homez Mouse anti-Human, Clone: PCRP-HOMEZ-1B5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30725820UG
Description
Homez Monoclonal specifically detects Homez in Human samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Homez | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
homeobox and leucine zipper encoding, homeobox and leucine zipper protein Homez, KIAA1443Homeodomain leucine zipper-containing factor | |
Recombinant full-length human Homez protein | |
20 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry (Paraffin), Peptide Array | |
PCRP-HOMEZ-1B5 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
57594 | |
Store at 4°C. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction