Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diazyme Laboratories Homocysteine Enzymatic Assay
Click to view available options
Type:
Homocysteine Enzymatic Assay
Quantity:
1 assay
Includes:
2 reagent assay
Description
- No significant interference from lipemic and hemolytic substances
- Eliminates carry over interference with iron or lipase reagents
- Assay not interfered by endogenous cystathionine
- For in vitro quantitative determination of total L-homocysteine in serum or plasma
- Assists in diagnosis and treatment of patients suspected of having hyperhomocysteinemia and homocystinuria
Specifications
Specifications
| Certifications/Compliance | 510(k) cleared, Health Canada registered |
| CE Marker | Yes |
| Type | Homocysteine Enzymatic Assay |
| For Use With (Equipment) | Beckman AU (400/600/640/680) analyzers |
| Includes | 2 reagent assay |
| Linearity | 3 to 50 μmol/L |
| Quantity | 1 assay |
| Form | Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction