Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Homosalate, Spectrum™ Chemical
SDP

Catalog No. 18603135
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle

H1187, 118-56-9, C16H22O3

Spectrum™ Chemical Homosalate is an organic compound most often used in sunscreens as a chemical UV filter. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 118-56-9
Molecular Formula C16H22O3
Molecular Weight (g/mol) 262.35
InChI Key WSSJONWNBBTCMG-UHFFFAOYNA-N
IUPAC Name 3,3,5-trimethylcyclohexyl 2-hydroxybenzoate
SMILES CC1CC(CC(C)(C)C1)OC(=O)C1=CC=CC=C1O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 97 to 103%
Grade Reagent
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.