Learn More
Honeywell Analytics™ GasAlert Extreme Single-Gas Detectors
Description
For any single gas hazard, choose from a wide range of available toxic gas models. TheGasAlert Extreme Single-Gas Detector provides a simple, low-cost option requiringminimal training. Its compact, lightweight design makes it versatile and comfortable.
- Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms on start up
- Secure and accurate history with datalogging with passcode protection
- Continuous LCD shows real-time gas concentrations
- Simple automatic calibration procedure
- Equipped with internal vibrating alarm for high noise areas
- Visual, vibrating, audible alarms (95 dB) - Low, High, TWA and STEL
- Replaceable 3V battery
- Water-resistant
- Bright wide-angled visual alarm bars
- Two-year field-replaceable battery (1.5 year life) and sensor
- Adjustable alarm setpoints
- Multilanguage display options
- Compatible with Sampler motorized pump
- Tests: Sensor integrity, circuitry, battery and audible/visual alarms on activation, battery
- Humidity Range: 15% to 90% for all but O2 (0% to 99%) and Cl2 (10% to 95%) sensors
- Size: 2.8 × 5.0 × 9.5cm (1.1 × 2.0 × 3.75 in.)
- Weight: 82g (2.9 oz.)
Warranty and Services
Two years including sensor, except Ammonia (one year)
Specifications
Specifications
| Certifications/Compliance | ATEX: CE Ex II 1 G Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, Class II, Division 1, Group E, F, G, IECEx: Exia IIC T4, INMETRO: BR-Exia IIC T4, EMC Directive 89/336/EEC |
| Depth (English) | 1.1 in. |
| Depth (Metric) | 2.8 cm |
| Components Measured | Chlorine |
| Type | Chlorine Detector |
| Height (English) | 3.75 in. |
| Height (Metric) | 9.52 cm |
| Humidity Range | 10 to 95% |
| Measurement Range | 0 to 50ppm |
| Temperature Range (English) | -4°F to +122°F |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.