Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Analytics™ GasAlert Extreme Single-Gas Detectors

For any single gas hazard, choose from a wide range of available toxic gas models. The GasAlert Extreme Single-Gas Detector provides a simple, low-cost option requiring minimal training. Its compact, lightweight design makes it versatile and comfortable.

Supplier:  Honeywell Analytics™ GAXTEDL

 View more versions of this product

Catalog No. 19-413-358


Only null left
Add to Cart

Description

Description

For any single gas hazard, choose from a wide range of available toxic gas models. TheGasAlert Extreme Single-Gas Detector provides a simple, low-cost option requiringminimal training. Its compact, lightweight design makes it versatile and comfortable.

  • Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms on start up
  • Secure and accurate history with datalogging with passcode protection
  • Continuous LCD shows real-time gas concentrations
  • Simple automatic calibration procedure
  • Equipped with internal vibrating alarm for high noise areas
  • Visual, vibrating, audible alarms (95 dB) - Low, High, TWA and STEL
  • Replaceable 3V battery
  • Water-resistant
  • Bright wide-angled visual alarm bars
  • Two-year field-replaceable battery (1.5 year life) and sensor
  • Adjustable alarm setpoints
  • Multilanguage display options 
  • Compatible with Sampler motorized pump
  • Tests: Sensor integrity, circuitry, battery and audible/visual alarms on activation, battery
  • Humidity Range: 15% to 90% for all but O2 (0% to 99%) and Cl2 (10% to 95%) sensors
  • Size: 2.8 × 5.0 × 9.5cm (1.1 × 2.0 × 3.75 in.)
  • Weight: 82g (2.9 oz.)

Warranty and Services

Two years including sensor, except Ammonia (one year)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATEX: CE Ex II 1 G Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, Class II, Division 1, Group E, F, G, IECEx: Exia IIC T4, INMETRO: BR-Exia IIC T4, EMC Directive 89/336/EEC
2.8 cm
Ethylene Oxide Detector
9.52 cm
0 to 100ppm
-20°C to +50°C
2.9 oz.
2 in.
1.1 in.
Ethylene Oxide
3.75 in.
15 to 90%
-4°F to +122°F
2 Years
83g
5.08 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.