Learn More
Honeywell Analytics™ GasAlert Extreme Single-Gas Detectors
For any single gas hazard, choose from a wide range of available toxic gas models. The GasAlert Extreme Single-Gas Detector provides a simple, low-cost option requiring minimal training. Its compact, lightweight design makes it versatile and comfortable.
Supplier: Honeywell Analytics™ GAXTPDL
Description
For any single gas hazard, choose from a wide range of available toxic gas models. TheGasAlert Extreme Single-Gas Detector provides a simple, low-cost option requiringminimal training. Its compact, lightweight design makes it versatile and comfortable.
- Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms on start up
- Secure and accurate history with datalogging with passcode protection
- Continuous LCD shows real-time gas concentrations
- Simple automatic calibration procedure
- Equipped with internal vibrating alarm for high noise areas
- Visual, vibrating, audible alarms (95 dB) - Low, High, TWA and STEL
- Replaceable 3V battery
- Water-resistant
- Bright wide-angled visual alarm bars
- Two-year field-replaceable battery (1.5 year life) and sensor
- Adjustable alarm setpoints
- Multilanguage display options
- Compatible with Sampler motorized pump
- Tests: Sensor integrity, circuitry, battery and audible/visual alarms on activation, battery
- Humidity Range: 15% to 90% for all but O2 (0% to 99%) and Cl2 (10% to 95%) sensors
- Size: 2.8 × 5.0 × 9.5cm (1.1 × 2.0 × 3.75 in.)
- Weight: 82g (2.9 oz.)
Warranty and Services
Two years including sensor, except Ammonia (one year)

Specifications
ATEX: CE Ex II 1 G Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, Class II, Division 1, Group E, F, G, IECEx: Exia IIC T4, INMETRO: BR-Exia IIC T4, EMC Directive 89/336/EEC | |
2.8 cm | |
3.75 in. | |
15 to 90% | |
-4°F to +122°F | |
2 Years | |
83g | |
5.08 cm |
1.1 in. | |
Phosphine | |
9.52 cm | |
0 to 5ppm | |
-20°C to +50°C | |
2.9 oz. | |
2 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.