Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Analytics™ GasAlert Extreme Single-Gas Detectors

Catalog No. 19413364
Encompass_Preferred
Change view
Click to view available options
Components Measured:
Ammonia
Chlorine
Chlorine Dioxide
Hydrogen Cyanide
Hydrogen Sulfide
Nitric Oxide
Nitrogen Dioxide
Ozone
Sulfur Dioxide
Measurement Range:
0 to 100ppm
0 to 1ppm
0 to 250ppm
0 to 30ppm
0 to 400ppm
0 to 500ppm
0 to 50ppm
0 to 99.9ppm
Humidity Range:
10 to 95%
15 to 90%
15 to 95%
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured Measurement Range Humidity Range
19-413-364 Sulfur Dioxide 0 to 100ppm 15 to 90%
19-413-360 Hydrogen Sulfide 0 to 100ppm 15 to 90%
19-413-359 Ozone 0 to 1ppm 15 to 90%
19-413-356 Chlorine 0 to 50ppm 10 to 95%
19-413-357 Nitrogen Dioxide 0 to 99.9ppm 15 to 95%
19-413-362 Nitric Oxide 0 to 250ppm 15 to 90%
19-413-368 Ammonia 0 to 400ppm 15 to 90%
19-413-367 Hydrogen Sulfide 0 to 500ppm 15 to 90%
19-413-365 Chlorine Dioxide 0 to 1ppm 15 to 95%
19-413-366 Hydrogen Cyanide 0 to 30ppm 15 to 95%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 19-413-364 Supplier Honeywell Analytics™ Supplier No. GAXTSDL

For any single gas hazard, choose from a wide range of available toxic gas models. The GasAlert Extreme Single-Gas Detector provides a simple, low-cost option requiring minimal training. Its compact, lightweight design makes it versatile and comfortable.

For any single gas hazard, choose from a wide range of available toxic gas models. TheGasAlert Extreme Single-Gas Detector provides a simple, low-cost option requiringminimal training. Its compact, lightweight design makes it versatile and comfortable.

  • Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms on start up
  • Secure and accurate history with datalogging with passcode protection
  • Continuous LCD shows real-time gas concentrations
  • Simple automatic calibration procedure
  • Equipped with internal vibrating alarm for high noise areas
  • Visual, vibrating, audible alarms (95 dB) - Low, High, TWA and STEL
  • Replaceable 3V battery
  • Water-resistant
  • Bright wide-angled visual alarm bars
  • Two-year field-replaceable battery (1.5 year life) and sensor
  • Adjustable alarm setpoints
  • Multilanguage display options 
  • Compatible with Sampler motorized pump
  • Tests: Sensor integrity, circuitry, battery and audible/visual alarms on activation, battery
  • Humidity Range: 15% to 90% for all but O2 (0% to 99%) and Cl2 (10% to 95%) sensors
  • Size: 2.8 × 5.0 × 9.5cm (1.1 × 2.0 × 3.75 in.)
  • Weight: 82g (2.9 oz.)

Warranty and Services

Two years including sensor, except Ammonia (one year)

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ATEX: CE Ex II 1 G Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, Class II, Division 1, Group E, F, G, IECEx: Exia IIC T4, INMETRO: BR-Exia IIC T4, EMC Directive 89/336/EEC
Depth (English) 1.1 in.
Depth (Metric) 2.8 cm
Components Measured Sulfur Dioxide
Type Sulfur Dioxide Detector
Height (English) 3.75 in.
Height (Metric) 9.52 cm
Humidity Range 15 to 90%
Measurement Range 0 to 100ppm
Temperature Range (English) -40°F to +122°F
Temperature Range (Metric) -40° to +50°C
Warranty 2 Years
Weight (English) 2.9 oz.
Weight (Metric) 83g
Width (English) 2 in.
Width (Metric) 5.08 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.