Learn More
Honeywell Analytics™ GasAlertMax XT II Single-Gas Detectors
Supplier:  Honeywell Analytics™ XT000MBNA
 View more versions of this product
Catalog No. 19-413-400
Only null left
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATEX: CE Ex II 1 G Ga Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, IECEx: Ga Exia IIC T4, INMETRO: BR-Exia IIC T4
2 in.
Carbon Monoxide
13.97 cm
66/67
1
-20°C to +50°C
11.1 oz.
3.2 in.
Black
5.08 cm
5.5 in.
10 to 100%
0 to 1000
-4°F to +122°F
2 Years
316g
8.1 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.