Learn More
Honeywell Analytics™ GasAlertQuattro Rechargable Single-Gas Detectors
Supplier: Honeywell Analytics™ QT00H0RYNA
Description
Specifications
ATEX: CE Ex II 1 G Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, IECEx: Exia IIC T4, INMETRO: BR-Exia IIC T4, EMC Directive 2004/108/EC | |
2 in. | |
Hydrogen Sulfide | |
8.12 cm | |
66/67 | |
0.1 | |
-20°C to +50°C | |
11.15 oz. | |
5.1 in. |
Yellow | |
5.08 cm | |
3.2 in. | |
10 to 100% | |
0 to 200 | |
-4°F to +122°F | |
2 Years | |
316g | |
12.95 cm |