Learn More
Honeywell Analytics™ Integral Motorized Pump Kit
For GasAlertMicro 5 Series Multi-Gas Detectors.
Supplier:  Honeywell Analytics™ M5PUMP
Catalog No. M5PUMPBP
Only null left
Add to Cart
Includes: Motorized pump cover, 30cm (1 ft.) sample probe with hydrophobic barrier (water trap) and particulate filters, 1.5m (5 ft.) sampling tubing, three spare particulate filters and one spare water trap filter
Description
Description
For GasAlertMicro 5 Series Multi-Gas Detectors.
  • With visible integrated yellow filter
  • For GasAlertMicro 5 Series Multi-Gas Detectors
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GasAlertMicro 5 Series Multi-Gas Detectors
Integral Motorized Pump Kit
Motorized pump cover, 30cm (1 ft.) sample probe with hydrophobic barrier (water trap) and particulate filters, 1.5m (5 ft.) sampling tubing, three spare particulate filters and one spare water trap filter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.