Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ NorthFlex Duro Task™ Textured Latex Coated Seamless Knit Gloves

Catalog No. 191500948 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-150-0948 X-Large
19-150-0945 Small
19-150-0946 Medium
19-150-0947 Large
19-150-0949 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-150-0948 Supplier Honeywell™ Supplier No. NF14/10XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Gloves feature a seamless knit shell and textured latex coating on the palm and fingertips for enhanced grip capabilities.

  • Rough latex coating enhances grip capabilities
  • Seamless cotton/polyester construction increases user comfort and finger sensitivity
  • Ideal for general purpose applications such as assembly work, construction, and warehousing

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Latex
Color Blue, Gray
Exterior Finish Fully Textured
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size X-Large
For Use With (Application) Automotive Assembly, Construction, and Warehousing
Glove Cuff Style Knit
Series NF14
Yarn Gauge 10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.