Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ NorthFlex Light Task Plus 5™ Cut-Resistant Gloves

Catalog No. 19999128A Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-999-128A Small
19-999-128B Medium
19-999-128C Large
19-999-128D X-Large
19-999-128E 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-999-128A Supplier Honeywell™ Supplier No. NFD20B/7S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Bi-polymer coated cut-resistant gloves is a blend that combines the best properties of nitrile and PU (polyurethane) to provide a surface that is mechanically tough and flexible.

  • Bi-polymer coating is a blend of nitrile and PU (polyurethane)
  • Comfortable, high performance cut-resistant
  • Seamless form fitting liner contains 'stretch-broken' Dynamo yarn that is cool and flexible
  • 13-gauge liner also contains glass fiber for enhanced cut resistance
  • Knit wrist

Specifications

Coating Material Bi-Polymer
Color Black
Cut Resistance Rating ANSI A5
Hand Specific
Material Dyneema Fiber
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Material handling, assembly work, sheet metal, glass
Glove Cuff Color Orange
Glove Cuff Style Knit
Model NFD20B/7S
Series NFD20B
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.