Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ SilverShield™ SSG Ink-Free Gloves

Catalog No. 19048511H
Click to view available options
Size:
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Unique five-layer fabric is resistant to over 280 different chemicals including alcohols, aliphatics, aromatice, chlorines, ketones and esters. Ink-free version is offered without any logo or printed information on the glove.

  • Unique five-layer construction provides chemical protection
  • Ideal for hospitals, agriculture, military and hazardous material handling
  • Easily worn under heavy-duty outer gloves
  • Use as a primary glove where there is minimal risk of tearing or puncturing
  • Resistant to over 280 different chemicals, including alcohols, aliphatics, aromatics, chlorines, ketones and esters
  • Does not contain latex or chemical accelerators
  • Ink-free version does not contain any printed information on the glove
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OSHA 29 CFR 1915.157
Certifications/Compliance (Europe) EN 420, EN 374-3
Color Silver
Hand Ambidextrous
Latex Free Yes
Material Polyethylene, EVA
Size 9
Thickness (English) 2.7 mil
Thickness (Metric) 0.06 mm
EN388 Standard EN 388
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Straight
Grip Design Smooth
Lining Unlined
Model SSG/9/IF
Series SilverShield™ SSG/IF
Length (English) 14.5 in.
Length (Metric) 37 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.