Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Junk Yard Dog™ Aramid Fiber/Leather Cut-Resistant Gloves

Catalog No. p-4365044
Click to view available options
Size:
Large
Glove Cuff Color:
Black
This item is not returnable. View return policy

Gloves are cut resistant - not cut proof. Extreme caution should be used around moving machinery.

Laundering gloves that have been in contact with toxic or corrosive chemicals may result in a chemical reaction, including fumes, that could cause personal injury, serious illness or death.

This item is not returnable. View return policy

Gloves are cut resistant - not cut proof. Extreme caution should be used around moving machinery.

Laundering gloves that have been in contact with toxic or corrosive chemicals may result in a chemical reaction, including fumes, that could cause personal injury, serious illness or death.

Aramid fiber gloves with premium split leather palm offer abrasion resistance, oil resistance, tear resistance and cut resistance.

  • Shell is made of 100% aramid fiber
  • Premium split leather coated
  • Cut, slash, heat and flame resistance
  • Full leather reinforced palm provide puncture and abrasion resistance
  • Available in three colors and sizes

TRUSTED_SUSTAINABILITY

Specifications

Coating Material Leather
Color Yellow, Gray
Cut Resistance Rating ANSI A4
Material Aramid Fiber, Leather
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) Construction, manufacturing, steel and metals
Gender Men's
Glove Cuff Color Black
Model KV20A-100-50
Series Junk Yard Dog
Yarn Gauge 7
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.