Learn More
Honeywell™ Kangaroo Leather Super Glove™ - Gauntlet Cuff
Catalog No. 19171019F
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
3D hand-shaped design with staggered layer seaming is ergonomically correct, helping to fight hand fatigue and reduce injuries. Digiroo™ High Grip technology on the palm and fingers ensures dexterity, grip, and tactility.
  • Air Spacer™ Thermal Architecture traps air for thermal protection without adding bulk to the glove
  • Black Digiroo™ Digital Kangaroo Grip on the palm and fingers for durability, grip, and tactility
  • Crosstech™ Moisture Barrier is stable and durable with high temperature resistance
  • Bubble-Flex Construction on the knuckles and fingers features dead-air spacer ridges for natural hand flexing
  • Top Grain Kangaroo Leather provides abrasion resistance and exceptional durability
  • Anthropomorphically correct 3D-style fights hand fatigue and reduces injury
Certifications
NFPA 1971-2007
TRUSTED_SUSTAINABILITY
Specifications
Back Material Top Grain Kangaroo Leather
Certifications/Compliance NFPA 1971-2007
Coated Area Palm, Fingers
Cuff Material Cowhide Leather
Color Blue
Hand Specific
Product Type Structural Firefighting Gloves
Size 2X-Large
Glove Cuff Color Black
Glove Cuff Style Gauntlet
Includes Crosstech™ Moisture Barrier
Liner Material Nomex™/Aramid Fiber
Series GL-SGKCG
Thumb Style Wing
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.