Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Nitri-Kote™ Nitrile Coated Cotton/Polyester Gloves

Catalog No. 191500921
Click to view available options
Size:
X-Small
Small
Medium
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

A unique coating process allows the glove to be fully coated for extended wear and breathability. The blended cotton/polyester shell reduces shrinking when laundered.

  • Ambidextrous design allows gloves to be replaced one at a time, rather than in pairs
  • Seamless construction increases comfort with no rubbing or chafing
  • Complies with USDA and FDA regulations (21 CFR) for use in food processing
  • Knit wrist prevents dirt and debris from entering the glove
TRUSTED_SUSTAINABILITY

Specifications

Coated Area Full Glove
Construction Seamless Knit
Coating Material Nitrile
Color Orange
Hand Ambidextrous
Material Polycotton
Size X-Large
For Use With (Application) Food Processing
Glove Cuff Style Knit
Series 78/1142
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.