Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ NorthFlex Neon™ Foam PVC Coated Hi-Vis Polyamide Gloves

Catalog No. 17805005 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-805-005 Small
17-805-008 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-805-005 Supplier Honeywell™ North™ Supplier No. NF11HVY/7S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ergonomic, seamless knit construction conforms to the natural shape of the hand for a comfortable fit. Hi-Vis yellow color increases visibility for tasks in low-light areas.

  • Hi-vis yellow color increases worker visibility in low-light areas
  • 15 gauge seamless liner is stretchy and form fitting for user comfort
  • Foam PVC coating enhances grip capabilities, especially in wet areas
  • Coating is soft and flexible to not compromise tactility or dexterity

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material PVC
Color Hi-vis Yellow
Hand Specific
Latex Free Yes
Material Nylon
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Construction, Municipal Services, and Transportation
Glove Cuff Style Knit
Series NF11HVY
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.