Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Perfect Fit™ Aramid Fiber Cut-Resistant Gloves, PVC Dot Coated
Click to view available options
Size:
Medium
Description
- Seamless knit, aramid fiber
- Protects against cut, slash and abrasion
- Not recommended for thermal heat protection
- Extremely durable and comfortable seamless construction
- Ambidextrous
- Rugged, high-quality brown dot coating on both sides to enhance grip

Specifications
Specifications
ANSI Abrasion Rating | ANSI 3 |
Coating Material | PVC Dots (2 Sides) |
Color | Yellow, Brown |
Cut Resistance Rating | ANSI A3 |
Hand | Ambidextrous |
Material | Aramid Fiber |
Product Type | Cut-resistant Gloves |
Size | Medium |
EN388 Standard | EN 388 |
For Use With (Application) | automotive and part manufacturer, building and construction, fishing, general industry, Glass industries, Metal Steel, mining and quarrying, ship building, Wood industries |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction