Learn More
Honeywell™ PowerCoat™ 660 Blue PVC Gloves
Catalog No. 19049927C Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-049-927C X-Large
19-049-927A Medium
19-049-927B Large
19-049-927D 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-049-927C Supplier Honeywell™ Supplier No. 660XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
PVC dipped coated gloves protect in applications where hands are immersed or splashed with hazardous liquids and chemicals.
  • Superior grip and dexterity, flexibility reduces hand fatigue and promotes increased productivity
  • Supple as natural latex, but has the chemical resistance of PVC
  • Ideal alternative for workers with latex allergies
  • Ideal for applications including glass handling, injection molding, plastic processing and more
Specifications
Color Blue
Latex Free Yes
Material PVC
Size X-Large
Thickness (English) 59 mil
Thickness (Metric) 1.50 mm
EN388 Standard EN 388
Glove Cuff Style Pinked
Grip Design Rough
Liner Material Cotton
Lining Lined
Model 660-XL
Series PowerCoat™ 660
Length (English) 12 in.
Length (Metric) 30.5 cm