Learn More
Honeywell™ PRO 5006 Structural Firefighting Boots, EEE Width
Superior combination of comfort and absorbency
Supplier:  Honeywell™ BT5006B0905
 View more versions of this product
Catalog No. 19-167-444
Only null left
Add to Cart
Description
Description
Quilted to 7.5 oz Kevlar™ felt thermal barrier and laminated to fully encapsulating Crosstech™ Footwear Fabric bootie. Crosstech™ moisture barrier provides superior liquid penetration resistance to blood, body fluids and NFPA tested common chemicals.
  • Military AB leather construction insulates and adds abrasion-, puncture-, water- and chemical-resistance
  • With heavy duty 0.75 in. reinforced straps mounted to the exterior of boot and anchored with a 2 × 1.5 in. shield-shaped base
  • External texon board shinguard protects and guards sensitive shin area of legs
  • Patented PowerToe™ cover resists toe abrasion and provides unparalled wear over the lifetime of the boots
  • Oblique toe safety cap provides additional room in height and width of boot
  • Fire/Ice PowerSole™ angled lugs provide substantial edging and gripping capabilities
  • Molded thermoplastic heel counter locks foot into the boot and anchors it in place
  • Patented PowerHeel™ cover protects heel area from abusive doffing of boots
  • Corrison-resistant steel ladder shank and stainless steel, full length, toe-to-heel puncture resistant bottom plate
Order Info
D and E widths also available.
Compliance
Meet NFPA 1971, 2007 edition requirements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
14 in.
Pro-V Liner
9EEE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.