Learn More
Honeywell™ PRO 5006 Structural Firefighting Boots, EEE Width
Superior combination of comfort and absorbency
$795.00
Catalog Number | Mfr. No. | Material | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Size | Price | Quantity & Availability | ||||
19-167-447
|
Honeywell™
BT5006B1055 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 10.5EEE |
Pair for $795.00
|
|
||||
19-167-446
|
Honeywell™
BT5006B1005 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 10EEE |
Pair for $795.00
|
|
||||
19-167-451
|
Honeywell™
BT5006B1255 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 12.5EEE |
Pair for $795.00
|
|
||||
19-167-448
|
Honeywell™
BT5006B1105 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 11EEE |
Pair for $795.00
|
|
||||
19-167-450
|
Honeywell™
BT5006B1205 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 12EEE |
Pair for $795.00
|
|
||||
19-167-452
|
Honeywell™
BT5006B1305 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 13EEE |
Pair for $795.00
|
|
||||
19-167-453
|
Honeywell™
BT5006B1405 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 14EEE |
Pair for $795.00
|
|
||||
19-167-454
|
Honeywell™
BT5006B1505 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 15EEE |
Pair for $795.00
|
|
||||
19-167-437
|
Honeywell™
BT5006B0555 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 5.5EEE |
Pair for $795.00
|
|
||||
19-167-455
|
Honeywell™
BT5006B1605 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 16EEE |
Pair for $795.00
|
|
||||
19-167-438
|
Honeywell™
BT5006B0605 |
Pro-V Liner | 6EEE |
Pair for $795.00
|
|
||||
19-167-439
|
Honeywell™
BT5006B0655 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 6.5EEE |
Pair for $795.00
|
|
||||
19-167-440
|
Honeywell™
BT5006B0705 |
Pro-V Liner | 7EEE |
Pair for $795.00
|
|
||||
19-167-441
|
Honeywell™
BT5006B0755 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 7.5EEE |
Pair for $795.00
|
|
||||
19-167-443
|
Honeywell™
BT5006B0855 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 8.5EEE |
Pair for $795.00
|
|
||||
19-167-442
|
Honeywell™
BT5006B0805 |
Pro-V Liner | 8EEE |
Pair for $795.00
|
|
||||
19-167-445
|
Honeywell™
BT5006B0955 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 9.5EEE |
Pair for $795.00
|
|
||||
19-167-444
|
Honeywell™
BT5006B0905 |
Pro-V Liner | 9EEE |
Pair for $795.00
|
|
||||
19-167-449
|
Honeywell™
BT5006B1155 |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | 11.5EEE |
Pair for $795.00
|
|
||||
Description
Quilted to 7.5 oz Kevlar™ felt thermal barrier and laminated to fully encapsulating Crosstech™ Footwear Fabric bootie. Crosstech™ moisture barrier provides superior liquid penetration resistance to blood, body fluids and NFPA tested common chemicals.
- Military AB leather construction insulates and adds abrasion-, puncture-, water- and chemical-resistance
- With heavy duty 0.75 in. reinforced straps mounted to the exterior of boot and anchored with a 2 × 1.5 in. shield-shaped base
- External texon board shinguard protects and guards sensitive shin area of legs
- Patented PowerToe™ cover resists toe abrasion and provides unparalled wear over the lifetime of the boots
- Oblique toe safety cap provides additional room in height and width of boot
- Fire/Ice PowerSole™ angled lugs provide substantial edging and gripping capabilities
- Molded thermoplastic heel counter locks foot into the boot and anchors it in place
- Patented PowerHeel™ cover protects heel area from abusive doffing of boots
- Corrison-resistant steel ladder shank and stainless steel, full length, toe-to-heel puncture resistant bottom plate
Order Info
D and E widths also available.
Compliance
Meet NFPA 1971, 2007 edition requirements
Specifications
14 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.