Learn More
Honeywell™ PRO 5007 Structural Firefighting Bunker Boots - D Width
Quilted to 7.5 oz. Kevlar™ felt thermal barrier and laminated to fully encapsulating Crosstech™ Footwear Fabric bootie
$808.50 - $1177.00
Specifications
Height (English) 14 in.
View More Specs
Products 21
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
18-821-416
View Documents
Honeywell™
BT50070402
4D
Pair for $1,177.00
Only null left
Add to Cart
18-821-417
View Documents
Honeywell™
BT50070452
4.5DC
Pair for $1,177.00
Only null left
Add to Cart
18-821-418
View Documents
Honeywell™
BT50070502
5D
Pair for $808.50
Only null left
Add to Cart
18-821-422
View Documents
Honeywell™
BT50070702
7D
Pair for $812.00
Only null left
Add to Cart
18-821-420
View Documents
Honeywell™
BT50070602
6DC
Pair for $812.00
Only null left
Add to Cart
18-821-421
View Documents
Honeywell™
BT50070652
6.5D
Pair for $812.00
Only null left
Add to Cart
18-821-423
View Documents
Honeywell™
BT50070752
7.5D
Pair for $812.00
Only null left
Add to Cart
18-821-419
View Documents
Honeywell™
BT50070552
5.5D
Pair for $808.50
Only null left
Add to Cart
18-821-424
View Documents
Honeywell™
BT50070802
8D
Pair for $812.00
Only null left
Add to Cart
18-821-425
View Documents
Honeywell™
BT50070852
8.5D
Pair for $812.00
Only null left
Add to Cart
18-821-426
View Documents
Honeywell™
BT50070902
9D
Pair for $812.00
Only null left
Add to Cart
18-821-427
View Documents
Honeywell™
BT50070952
9.5D
Pair for $812.00
Only null left
Add to Cart
18-821-428
View Documents
Honeywell™
BT50071002
10D
Pair for $812.00
Only null left
Add to Cart
18-821-431
View Documents
Honeywell™
BT50071152
11.5D
Pair for $812.00
Only null left
Add to Cart
18-821-429
View Documents
Honeywell™
BT50071052
10.5D
Pair for $812.00
Only null left
Add to Cart
18-821-430
View Documents
Honeywell™
BT50071102
11D
Pair for $812.00
Only null left
Add to Cart
18-821-432
View Documents
Honeywell™
BT50071202
12D
Pair for $812.00
Only null left
Add to Cart
18-821-434
View Documents
Honeywell™
BT50071302
13D
Pair for $812.00
Only null left
Add to Cart
18-821-433
View Documents
Honeywell™
BT50071252
12.5D
Pair for $812.00
Only null left
Add to Cart
18-821-435
View Documents
Honeywell™
BT50071402
14D
Pair for $812.00
Only null left
Add to Cart
18-821-436
View Documents
Honeywell™
BT50071502
15D
Pair for $812.00
Only null left
Add to Cart
Description
Description
  • Crosstech™ moisture barrier provides superior liquid penetration resistance to blood, body fluids and NFPA tested common chemicals
  • Military AB leather construction insulates and adds abrasion-, puncture-, water- and chemical-resistance
  • With heavy duty 0.75 in. reinforced straps mounted to the exterior of boot and anchored with a 2 × 1.5 in. shield-shaped base
  • External texon board shinguard protects and guards sensitive shin area of legs
  • Patented PowerToe™ cover resists toe abrasion and provides unparalled wear over the lifetime of the boots
  • Oblique toe safety cap provides additional room in height and width of boot
  • Fire/Ice PowerSole™ angled lugs provide substantial edging and gripping capabilities
  • Molded thermoplastic heel counter locks foot into the boot and anchors it in place
  • Patented PowerHeel™ cover protects heel area from abusive doffing of boots
  • Corrison-resistant steel ladder shank and stainless steel, full length, toe-to-heel puncture resistant bottom plate
Compliance
Meet NFPA 1971, 2007 edition requirements
Specifications
Specifications
14 in.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.