Learn More
Honeywell™ PRO 5050 Struximity Leather Boots, Berry-Compliant, Wide
Lightweight, comfortable, high-speed, wide-width leather pull-on bunker boots dual certified for both Structural and Proximity (ARFF) firefighting
Supplier:  Honeywell™ BT5050B0853
 View more versions of this product
Catalog No. 19-813-928
Only null left
Add to Cart
Description
Description
Made of high performance fire resistant and waterproof leather and Goodyear welt made to withstand the harsh proximity environment
  • Athletic construction delivers unencumbered performance and agility
  • Full height bootie combines an aluminized radiant shield, 7.5 oz Kevlar thermal felt quilted to Cambrelle™ liner and laminated to Crosstech™ Moisture barrier fabric for protection from heat, fluids, and internal abrasion
  • Specially designed profiled boot shaft is large enough to accommodate large calves while still fitting comfortably under proximity pants
  • Vibram™ lug replaceable soles with Fire and Ice compound are cemented and security stitched using Goodyear welt construction for superior durability protection
  • Fire and Ice sole compound improves winter traction on ice and snow
  • Steel bottom plate and shank offer protection from puncture and dangerous twisting
  • Improved heel pocket and comfortable molded insert defend against heel slippage
  • APT Barcode — Departmental NFPA 1851 Tracking
  • Certified to NFPA 1971, current edition for Proximity and Structural Fire Fighting
  • Weight: 6.6 pounds
  • Wide width
  • Union made in USA. All components are Berry Amendment compliant
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cambrelle
Leather, CrossTech, Dupont Kevlar Aramid Fiber
8.5E
6.6 lb.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.