Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Safety Products™ Saline Wall Station
Click to view available options
Product Type:
Double Eye Wall Station
Single Eye Wall Station
Capacity (English):
16 oz.
32 oz.
Description
The Honewell Saline Wall Station is easily mounted in hazardous areas to provide immediate relief in case of eye injury.
Choose from 16 or 32 oz. (473 or 946mL) bottles.
- Highly visible, with full emergency-use instructions on front
- Made from sturdy, high-impact plastic
- Bottles are single-use only with snap-off tops for quick and easy access
- Bottles have extended flow nozzles; 32 oz. bottles have integral grip for ease of use in an emergency
Order Info
Refill bottles sold separately. See Cat. Nos. 19-003-030 and -033.
Compliance
Does not meet the criteria of plumbed or self-contained eyewash equipment.

Specifications
Specifications
Capacity (English) | 32 oz. |
Capacity (Metric) | 946 mL |
Certifications/Compliance | FDA |
For Use With (Application) | Flushing Nuisance Particles Such as Pollen, Dust, Saw Dust, Smoke and More |
For Use With (Equipment) | For Refill Bottles, See 19-003-030. |
Includes | Sperian Saline Buffered, Saline solution superior to tap Water |
Material | Polyethylene |
Product Type | Single Eye Wall Station |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction