Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Tuff-Coat™ Aramid Fiber Cut-Resistant Gloves, Latex Coated

Catalog No. 19175449
Click to view available options
Size:
Small
Large
X-Large
Glove Cuff Color:
Red
White
Yellow
This item is not returnable. View return policy

Catalog No. 19-175-449

Supplier: Honeywell™ KV200S

This item is not returnable. View return policy

Aramid fiber seamless knit liners with durable quality coatings are ideal for any task requiring cut protection.

Ideal for construction, forestry, municipal services, and transportation

  • Standard weight, 7-gauge Kevlar™ seamless knit shell
  • Cut and slash protection
  • Natural rubber palm and fingertip coating
  • Rubber coating provides excellent grip and flexibility
  • This product contains natural rubber

TRUSTED_SUSTAINABILITY

Specifications

Coating Material Natural Rubber
Color Yellow
Cut Resistance Rating ANSI A2
Latex Free No
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) construction, forestry, municipal services and transportation
Glove Cuff Color Red
Glove Cuff Style Knit
Hem Color Red
Series Tuff-Coat
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.