Honeywell Tuff-Coat™ Dipped Gloves

Feature seamless knit liners and durable quality coatings

Manufacturer: Honeywell 200XL

 View more versions of this product

Catalog No. 19-130-3903D

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit
    For Use With (Application) Provides extra protection, inhibit liquid penetration
    Liner Material Cotton, Polyester
    Cleaning Procedure Withstands commercial laundry room detergents (up to pH 14) and warm water temperatures up to 120°F. Lower heat settings up to 180°F are suggested for drying as extreme heat may cause shrinking and degradation.
    Exterior Finish Rough
    Glove Cuff Color White
    Material Natural Rubber
    Size X-Large
    Coated Area Palm, finger
    Color Gray, black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit